Assay Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009255.4 | NP_033281.1 | 7 - 8 | 1249 | 109 | |
| GenBank mRNA | AK045954.1 | - | 7 - 8 | 1221 | 109 | |
| AK047856.1 | - | 7 - 8 | 1221 | 109 | ||
| AK051332.1 | - | 7 - 8 | 1221 | 109 | ||
| AK159687.1 | - | 7 - 8 | 1221 | 109 | ||
| BC010675.1 | - | 7 - 8 | 1199 | 3595407 | 109 | |
| CT010311.1 | - | 6 - 7 | 1072 | 109 | ||
| X70296.1 | - | 7 - 8 | 109 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Serpine2
Genomic Map