Assay Details
| Gene Symbol: 4930519K11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK019684.1 | - | 2 - 3 | 302 | 119 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
potassium large conductance calcium-activated channel, subfamily M, alpha member 1 |
Kcnma1 |
5730414M22Rik, BKCa, MaxiK, Slo, Slo1, mSlo, mSlo1 |
-344.0 | Anti-sense | Chr. 14 - 23292291 - 24004951 on NCBI Build GRCm38 |
| Downstream | microRNA 7210 | Mir7210 |
mir-7210, mmu-mir-7210 |
83.0kb | Sense | Chr. 14 - 24088877 - 24088930 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

4930519K11Rik
Genomic Map