Assay Details
| Gene Symbol: Gm5092 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030245.1 | - | 1 - 2 | 31 | 85 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | zinc finger protein 160 | Zfp160 |
6720480D16Rik, F11, HKr18, HZF5, KR18, mKIAA1611, mszf47 |
-62.4kb | Sense | Chr. 17 - 21008941 - 21028857 on NCBI Build GRCm38 |
| Downstream | vomeronasal 1 receptor 234 | Vmn1r234 | V1rf1 | 133.8kb | Sense | Chr. 17 - 21228826 - 21229815 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm5092
Genomic Map