| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix HT MG-430 PM Array Plate |
1439277_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439277_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1439277_PM_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR1700020896.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020896.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020896.mm.1 |
NR_045913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020896.mm.1 |
NR_045913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020896.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020894.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020894.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020894.mm.1 |
NR_045913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020894.mm.1 |
NR_045913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020894.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020901.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020901.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020901.mm.1 |
NR_045913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020901.mm.1 |
NR_045913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020901.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020895.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020895.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020895.mm.1 |
NR_045913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020895.mm.1 |
NR_045913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020895.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020900.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020900.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020900.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020898.mm.1 |
NR_045912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020898.mm.1 |
NR_045912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020898.mm.1 |
NR_045913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020898.mm.1 |
NR_045913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020898.mm.1 |
TC1700002212.mm.1 |
MTA TC |
| Mouse Expression Set 430 |
1439277_PM_at |
- |
- |
| Mouse Expression Set 430 |
1439277_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439277_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1439277_at |
- |
- |