Assay Details
| Gene Symbol: Nkx2-2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_030769.2 | 1 - 2 | 275 | 77 | ||
| GenBank mRNA | AK045921.1 | - | 1 - 2 | 276 | 77 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | NK2 homeobox 4 | Nkx2-4 |
1700001P03Rik, Nkx-2.4, tinman |
-98.6kb | Anti-sense | Chr. 2 - 147083337 - 147085439 on NCBI Build GRCm38 |
| Downstream | expressed sequence AI646519 | AI646519 |
Aspax1, OTTMUSG00000015701 |
75.1kb | Anti-sense | Chr. 2 - 147361541 - 147362793 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Nkx2-2os
Genomic Map