Assay Details
| Gene Symbol: Metap1d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025633.3 | NP_079909.1 | 5 - 6 | 559 | 81 | |
| GenBank mRNA | AJ414378.1 | - | 2 - 3 | 422 | 81 | |
| AK005241.1 | - | 5 - 6 | 556 | 81 | ||
| AK010067.1 | - | 5 - 6 | 559 | 81 | ||
| AK043123.1 | - | 2 - 3 | 1345 | 81 | ||
| BC051534.1 | - | 2 - 3 | 433 | 3466745 | 81 | |
| BC060739.1 | - | 4 - 5 | 2067 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Metap1d
Genomic Map