Assay Details
| Gene Symbol: Slc25a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172436.3 | NP_766024.1 | 16 - 17 | 1769 | 58 | |
| GenBank mRNA | AK030595.1 | - | 16 - 17 | 1799 | 58 | |
| AK083156.1 | - | 16 - 17 | 1770 | 58 | ||
| AK144958.1 | - | 16 - 17 | 1804 | 58 | ||
| AK147948.1 | - | 16 - 17 | 1785 | 58 | ||
| BC013845.1 | - | 16 - 17 | 1748 | 58 | ||
| BC024952.1 | - | 16 - 17 | 1962 | 58 | ||
| BC060505.1 | - | 16 - 17 | 1756 | 6311836 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a12
Genomic Map