Assay Details
| Gene Symbol: Tomm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164729.1 | NP_001158201.1 | 1 - 2 | 491 | 59 | |
| NM_025365.3 | NP_079641.1 | 1 - 2 | 491 | 59 | ||
| GenBank mRNA | AK003291.1 | - | 1 - 2 | 153 | 59 | |
| AK010470.1 | - | 1 - 2 | 203 | 59 | ||
| AK019199.1 | - | 1 - 2 | 153 | 59 | ||
| BC021590.1 | - | 1 - 2 | 496 | 5068609 | 59 | |
| BC037589.1 | - | 1 - 2 | 191 | 5007624 | 59 | |
| CA319049.1 | - | 1 - 2 | 140 | 59 | ||
| CA321392.1 | - | 1 - 2 | 75 | 59 | ||
More Information
Set Membership: |
|

Tomm6
Genomic Map