Assay Details
| Gene Symbol: Trerf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097623.1 | NP_001091092.1 | 13 - 14 | 3301 | 73 | |
| NM_172622.2 | NP_766210.1 | 10 - 11 | 3078 | 73 | ||
| GenBank mRNA | AK035176.1 | - | 8 - 9 | 2850 | 73 | |
| AK046822.1 | - | 8 - 9 | 2719 | 73 | ||
| AK154542.1 | - | 11 - 12 | 2988 | 73 | ||
| AK155404.1 | - | 11 - 12 | 2985 | 73 | ||
| AY081774.1 | - | 9 - 10 | 2988 | 73 | ||
| BC059215.1 | - | 13 - 14 | 3301 | 6412734 | 73 | |
| BC108416.1 | - | 8 - 9 | 2761 | 5697521 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trerf1
Genomic Map