Assay Details
| Gene Symbol: Trerf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097623.1 | NP_001091092.1 | 15 - 16 | 3462 | 70 | |
| NM_172622.2 | NP_766210.1 | 12 - 13 | 3239 | 70 | ||
| GenBank mRNA | AK046822.1 | - | 10 - 11 | 2880 | 70 | |
| BC051645.1 | - | 1 - 2 | 82 | 70 | ||
| BC059215.1 | - | 15 - 16 | 3462 | 6412734 | 70 | |
| BC108416.1 | - | 10 - 11 | 2922 | 5697521 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trerf1
Genomic Map