Assay Details
| Gene Symbol: Prph2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008938.1 | NP_032964.1 | 2 - 3 | 1045 | 76 | |
| GenBank mRNA | AK136280.1 | - | 2 - 3 | 1072 | 76 | |
| BC028958.1 | - | 2 - 3 | 1040 | 5363367 | 76 | |
| BC085266.1 | - | 2 - 3 | 1044 | 6490224 | 76 | |
| X14770.1 | - | 2 - 3 | 1045 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028094 | X14770.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028094 | BC026484.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028094 | BC028958.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028094 | BC085266.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2028094 | NM_008938.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729679 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284991 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 891758 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 449236 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666688 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 766580 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730996 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 5526 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76193 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142911 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P107227 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | X14770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | X14770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028094 | X14770.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028094 | BC026484.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028094 | BC028958.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028094 | BC085266.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2028094 | NM_008938.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209479 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209476 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209471 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209487 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209466 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209495 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209502 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209469 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209484 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209497 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209501 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209492 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209491 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209488 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209481 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209485 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209473 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209500 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209483 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31209493 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08791114 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142911 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P107227 | - | - |
| Whole Mouse Genome G4122A | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | BC026477.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | X14770.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | BC026484.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | NM_008938.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P320340 | AK136280.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P104484 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142911 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P107227 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13530769 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13530768 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13530774 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13155910 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209518 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209554 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209500 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209484 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209492 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209449 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209511 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209479 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209538 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209560 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209568 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209491 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209506 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209482 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209553 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209497 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209466 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209473 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209523 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209584 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209451 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209540 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209528 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209483 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209542 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209507 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209471 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209565 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209510 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209485 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209458 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209488 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209548 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209574 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209575 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209544 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209493 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209546 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209551 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209446 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209476 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209512 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209460 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209558 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31209545 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13530762 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209530 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13530770 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209513 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209479 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209476 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209507 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096148 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13435645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209488 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209462 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209582 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13064974 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209473 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13529701 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209484 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209458 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209482 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13148201 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209533 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209466 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209516 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13439576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209584 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209474 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121199 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13154297 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13096141 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13121194 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209512 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209523 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13435644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209463 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13212715 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209469 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209493 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13529700 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209471 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209500 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209460 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209457 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209483 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209464 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209497 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31209485 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10445637 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445635 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445636 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10445634 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420511_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10445633 | - | - |
| Mouse Genome 430 2.0 Array | 1420511_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420511_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19133_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19133_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700008666.mm.1 | NM_008938.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008666.mm.1 | NM_008938.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008666.mm.1 | TC1700000905.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700008665.mm.1 | NM_008938.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008665.mm.1 | NM_008938.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700008665.mm.1 | TC1700000905.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420511_PM_at | - | - |
| Mouse Expression Set 430 | 1420511_at | - | - |
| Murine Genome U74v2 Set | 101169_at | - | - |
| Murine Genome U74v2 Set | 101169_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420511_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420511_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420511_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1420511_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 445804 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5550152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5550152 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 102480605 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005550152 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1241602 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2553522 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1241602 | - | - |
| mouse-6 v1.1 (Illumina) | scl50667.3_448-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 102480605 | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5550152 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl50667.3_448-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5550152 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102480605 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1241602 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223554 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241602 | - | - |

Prph2
Genomic Map