| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700020855.mm.1 |
NM_016859.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020855.mm.1 |
NM_016859.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020855.mm.1 |
TC1700002208.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020854.mm.1 |
NM_016859.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020854.mm.1 |
NM_016859.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020854.mm.1 |
TC1700002208.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020856.mm.1 |
NM_016859.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020856.mm.1 |
NM_016859.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020856.mm.1 |
TC1700002208.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020853.mm.1 |
NM_016859.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020853.mm.1 |
NM_016859.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020853.mm.1 |
TC1700002208.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700020857.mm.1 |
NM_016859.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020857.mm.1 |
NM_016859.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700020857.mm.1 |
TC1700002208.mm.1 |
MTA TC |