Assay Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019657.4 | NP_062631.1 | 3 - 4 | 318 | 76 | |
| GenBank mRNA | AF064635.1 | - | 3 - 4 | 297 | 76 | |
| AK078841.1 | - | 3 - 4 | 319 | 76 | ||
| AK081869.1 | - | 3 - 4 | 312 | 76 | ||
| AK082715.1 | - | 3 - 4 | 312 | 76 | ||
| AK088521.1 | - | 3 - 4 | 312 | 76 | ||
| AK150849.1 | - | 3 - 4 | 313 | 76 | ||
| AK152489.1 | - | 3 - 4 | 313 | 76 | ||
| AK166027.1 | - | 3 - 4 | 312 | 76 | ||
| AK167908.1 | - | 3 - 4 | 312 | 76 | ||
| BC010749.1 | - | 3 - 4 | 294 | 76 | ||
| BC090659.1 | - | 3 - 4 | 288 | 30675153 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b12
Genomic Map