Assay Details
| Gene Symbol: Hsd17b12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019657.4 | NP_062631.1 | 8 - 9 | 658 | 70 | |
| GenBank mRNA | AF064635.1 | - | 8 - 9 | 637 | 70 | |
| AK012103.1 | - | 1 - 2 | 72 | 70 | ||
| AK078841.1 | - | 8 - 9 | 659 | 70 | ||
| AK081869.1 | - | 8 - 9 | 652 | 70 | ||
| AK082715.1 | - | 8 - 9 | 652 | 70 | ||
| AK088521.1 | - | 8 - 9 | 652 | 70 | ||
| AK150849.1 | - | 8 - 9 | 653 | 70 | ||
| AK152489.1 | - | 8 - 9 | 653 | 70 | ||
| AK166027.1 | - | 8 - 9 | 652 | 70 | ||
| AK167908.1 | - | 8 - 9 | 652 | 70 | ||
| AK188070.1 | - | 3 - 4 | 135 | 70 | ||
| BC010749.1 | - | 8 - 9 | 633 | 70 | ||
| BC037620.1 | - | 7 - 8 | 575 | 5375831 | 70 | |
| BC090659.1 | - | 8 - 9 | 628 | 30675153 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hsd17b12
Genomic Map