Assay Details
| Gene Symbol: Cyb5r2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205227.1 | NP_001192156.1 | 2 - 3 | 198 | 57 | |
| NM_177216.4 | NP_796190.1 | 2 - 3 | 198 | 57 | ||
| GenBank mRNA | AK085272.1 | - | 2 - 3 | 163 | 57 | |
| AK147952.1 | - | 2 - 3 | 199 | 57 | ||
| BC107238.1 | - | 2 - 3 | 142 | 40052160 | 57 | |
| BC107239.1 | - | 2 - 3 | 142 | 40052161 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cyb5r2
Genomic Map