Assay Details
| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027192.2 | NP_081468.1 | 3 - 4 | 674 | 76 | |
| GenBank mRNA | AK010374.1 | - | 1 - 2 | 50 | 76 | |
| BC018513.1 | - | 3 - 4 | 677 | 3597859 | 76 | |
| BC024414.1 | - | 3 - 4 | 646 | 4973810 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttl
Genomic Map