Assay Details
| Gene Symbol: Ttl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027192.2 | NP_081468.1 | 4 - 5 | 807 | 87 | |
| GenBank mRNA | AK010374.1 | - | 2 - 3 | 183 | 87 | |
| BC018513.1 | - | 4 - 5 | 810 | 3597859 | 87 | |
| BC024414.1 | - | 4 - 5 | 779 | 4973810 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ttl
Genomic Map