Assay Details
| Gene Symbol: Sult1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018751.2 | NP_061221.2 | 4 - 5 | 542 | 79 | |
| GenBank mRNA | BC045149.1 | - | 4 - 5 | 542 | 6308175 | 79 |
More Information
Updated Mapping Information
Set Membership: |
|

Sult1c1
Genomic Map