Assay Details
| Gene Symbol: Sult1c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018751.2 | NP_061221.2 | 6 - 7 | 762 | 150 | |
| GenBank mRNA | AF033653.1 | - | 6 - 7 | 773 | 150 | |
| BC045149.1 | - | 6 - 7 | 762 | 6308175 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfur compound metabolic process
metabolic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
alcohol sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
sulfur compound metabolic process
metabolic process
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
alcohol sulfotransferase activity
aryl sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987740 | BC045149.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1987740 | NM_018751.2 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135387 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P121639 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987740 | BC045149.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1987740 | NM_018751.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P519811 | AF033653.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245972 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245970 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245939 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245967 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245946 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245963 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245929 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245960 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245931 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245941 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245927 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245964 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245935 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245933 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245956 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245966 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245968 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245942 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245944 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245959 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245952 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245954 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245934 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245962 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245936 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245940 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245950 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245948 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245961 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P121639 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08806922 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08806875 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135387 | - | - |
| Whole Mouse Genome G4122A | A_51_P519811 | AF033653.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P112617 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P121639 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125862 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125855 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125860 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125856 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13125854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245881 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245798 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245941 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245793 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245815 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245885 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245810 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245789 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245843 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245773 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245939 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245886 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245877 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245901 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245891 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245970 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245905 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245925 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245903 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245933 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245852 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245837 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245868 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245817 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245805 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245845 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245856 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245892 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245889 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245913 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245989 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245960 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245964 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245874 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245841 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245973 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245911 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245944 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245871 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245767 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245865 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245872 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245975 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245862 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245935 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245858 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245819 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245898 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245808 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245875 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245896 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245870 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245895 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245880 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245860 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245968 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245888 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245867 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245839 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245813 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245990 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245802 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245882 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245854 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245982 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245927 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245922 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245942 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245879 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245784 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245840 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245934 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245941 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245946 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245922 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245930 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245950 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245977 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13180245 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13180246 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245948 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13144708 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13335794 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245942 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245940 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245953 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245960 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245976 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13459441 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245926 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245917 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13317079 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245982 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245944 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422724 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13422733 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245925 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245951 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13302028 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245929 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245954 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245975 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245939 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245921 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245918 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245973 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451834 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451835 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451831 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451832 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451829 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451830 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451833 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1420470_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451828 | - | - |
| Mouse Genome 430 2.0 Array | 1420470_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1420470_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20888_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 20888_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700021431.mm.1 | NM_018751.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021431.mm.1 | NM_018751.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021431.mm.1 | TC1700002292.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700021430.mm.1 | NM_018751.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021430.mm.1 | NM_018751.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021430.mm.1 | TC1700002292.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1420470_at | - | - |
| Mouse Expression Set 430 | 1420470_PM_at | - | - |
| Murine Genome U74v2 Set | 96421_at | - | - |
| Murine Genome U74v2 Set | 96421_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420470_at | - | - |
| Mouse Genome 430A 2.0 Array | 1420470_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 585754 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 1740348 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 1740348 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0001740348 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2713826 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2713826 | - | - |
| mouse-6 v1.1 (Illumina) | scl49792.8.1_29-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 1740348 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49792.8.1_29-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1740348 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2713826 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2713826 | - | - |

Sult1c1
Genomic Map