Assay Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026935.4 | NP_081211.3 | 4 - 5 | 580 | 81 | |
| GenBank mRNA | AK007384.1 | - | 4 - 5 | 440 | 81 | |
| AK144336.1 | - | 4 - 5 | 520 | 81 | ||
| AY005469.1 | - | 4 - 5 | 580 | 81 | ||
| BC022665.1 | - | 4 - 5 | 573 | 4223678 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sult1c2
Genomic Map