Assay Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026935.4 | NP_081211.3 | 7 - 8 | 989 | 116 | |
| GenBank mRNA | AK007384.1 | - | 7 - 8 | 849 | 116 | |
| AK144336.1 | - | 7 - 8 | 929 | 116 | ||
| AY005469.1 | - | 7 - 8 | 989 | 116 | ||
| BC022665.1 | - | 7 - 8 | 982 | 4223678 | 116 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
sulfation
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
sulfation
N-acetylglucosamine 6-O-sulfotransferase activity
N-acetylgalactosamine 4-O-sulfotransferase activity
heparan sulfate 2-O-sulfotransferase activity
sulfotransferase activity
HNK-1 sulfotransferase activity
transferase activity
heparan sulfate 6-O-sulfotransferase activity
trans-9R,10R-dihydrodiolphenanthrene sulfotransferase activity
1-phenanthrol sulfotransferase activity
3-phenanthrol sulfotransferase activity
4-phenanthrol sulfotransferase activity
trans-3,4-dihydrodiolphenanthrene sulfotransferase activity
9-phenanthrol sulfotransferase activity
2-phenanthrol sulfotransferase activity
phenanthrol sulfotransferase activity
1-hydroxypyrene sulfotransferase activity
galactose 3-O-sulfotransferase activity
proteoglycan sulfotransferase activity
cholesterol sulfotransferase activity
hydroxyjasmonate sulfotransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 337122 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 947259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 627530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 435087 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649032 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322415 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649186 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 431500 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 562734 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 159493 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 10839 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102820 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132256 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245421 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245423 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245408 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245406 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245426 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245409 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245410 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245424 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245414 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245416 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245413 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31245419 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08806690 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102820 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132256 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08806677 | - | - |
| Whole Mouse Genome G4122A | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | AY005469.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | BC022665.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | NM_026935.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | AK007384.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P353895 | AK144336.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P133006 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P08806677 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102820 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132256 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13397445 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13326669 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13326668 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13397441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245442 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245416 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245444 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245408 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245402 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245356 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245364 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245399 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245441 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245387 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245391 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245376 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245439 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245431 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245360 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245372 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245339 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245423 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245406 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245397 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245413 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245366 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245381 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245404 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245330 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245421 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245336 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245419 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245386 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245375 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245435 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245400 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245348 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245385 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245328 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245363 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245338 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31245358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245427 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245410 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13299759 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245409 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245397 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245404 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245391 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139755 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13326666 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245400 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13509229 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245405 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245416 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245428 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245423 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245395 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13139756 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245396 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245421 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13324552 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13299761 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245394 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245408 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31245413 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10451821 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451825 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451823 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451819 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451820 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451827 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451822 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451824 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10451826 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449409_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10451818 | - | - |
| Mouse Genome 430 2.0 Array | 1449409_at | - | - |
| Mouse Genome 430 2.0 Array | 1449409_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69083_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69083_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700021412.mm.1 | NM_026935.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021412.mm.1 | NM_026935.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021412.mm.1 | TC1700002287.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700021411.mm.1 | NM_026935.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021411.mm.1 | NM_026935.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700021411.mm.1 | TC1700002287.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449409_PM_at | - | - |
| Mouse Expression Set 430 | 1449409_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449409_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449409_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449409_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449409_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 441362 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 5910538 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 5910538 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0005910538 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1235747 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1235747 | - | - |
| mouse-6 v1.1 (Illumina) | scl49794.7.1_23-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 5910538 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49794.7.1_23-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5910538 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2612973 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2612973 | - | - |

Sult1c2
Genomic Map