Assay Details
| Gene Symbol: Sgol1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028232.2 | NP_082508.1 | 2 - 3 | 240 | 136 | |
| GenBank mRNA | AB193067.1 | - | 1 - 2 | 148 | 136 | |
| AB193068.1 | - | 1 - 2 | 148 | 136 | ||
| AK014357.1 | - | 2 - 3 | 234 | 136 | ||
| AK154188.1 | - | 2 - 3 | 259 | 136 | ||
| AK196456.1 | - | 2 - 3 | 184 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB193067.1,AB193068.1,AK014357.1,AK154188.1,NM_028232.2 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sgol1
Genomic Map