Assay Details
| Gene Symbol: Serpine2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009255.4 | NP_033281.1 | 2 - 3 | 438 | 102 | |
| GenBank mRNA | AK045954.1 | - | 2 - 3 | 410 | 102 | |
| AK047856.1 | - | 2 - 3 | 410 | 102 | ||
| AK051332.1 | - | 2 - 3 | 410 | 102 | ||
| AK159687.1 | - | 2 - 3 | 410 | 102 | ||
| AK160700.1 | - | 2 - 3 | 410 | 102 | ||
| AK164270.1 | - | 2 - 3 | 410 | 102 | ||
| BC010675.1 | - | 2 - 3 | 388 | 3595407 | 102 | |
| CT010311.1 | - | 1 - 2 | 261 | 102 | ||
| X70296.1 | - | 2 - 3 | 409 | 102 | ||
More Information
Set Membership: |
|

Serpine2
Genomic Map