Assay Details
| Gene Symbol: Mrpl44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081210.1 | NP_001074679.1 | 1 - 2 | 291 | 71 | |
| GenBank mRNA | AK019986.1 | - | 1 - 2 | 194 | 71 | |
| AK146024.1 | - | 1 - 2 | 103 | 71 | ||
| AK150537.1 | - | 1 - 2 | 53 | 71 | ||
| AK151854.1 | - | 1 - 2 | 53 | 71 | ||
| AK156814.1 | - | 1 - 2 | 206 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mrpl44
Genomic Map