Assay Details
| Gene Symbol: Ears2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014324.2 | - | 1223 | 99 | ||
| AK076043.1 | - | 6 - 7 | 1217 | 99 | ||
| AK167506.1 | - | 6 - 7 | 1235 | 99 | ||
| AK169225.1 | - | 6 - 7 | 1224 | 99 | ||
| BC025907.1 | - | 6 - 7 | 1241 | 5064949 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ears2
Genomic Map