Assay Details
| Gene Symbol: Ears2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK014324.2 | - | 1355 | 67 | ||
| AK076043.1 | - | 7 - 8 | 1349 | 67 | ||
| AK167506.1 | - | 7 - 8 | 1367 | 67 | ||
| AK169225.1 | - | 7 - 8 | 1356 | 67 | ||
| BC025907.1 | - | 7 - 8 | 1373 | 5064949 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ears2
Genomic Map