Assay Details
| Gene Symbol: Nr4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001139509.1 | NP_001132981.1 | 3 - 4 | 1162 | 92 | |
| NM_013613.2 | NP_038641.1 | 3 - 4 | 1308 | 92 | ||
| GenBank mRNA | AB014889.1 | - | 3 - 4 | 901 | 92 | |
| AK087495.1 | - | 4 - 5 | 1186 | 92 | ||
| AK158607.1 | - | 3 - 4 | 1309 | 92 | ||
| BC137715.1 | - | 3 - 4 | 1620 | 8860734 | 92 | |
| BC144854.1 | - | 3 - 4 | 1836 | 92 | ||
| S53744.1 | - | 3 - 4 | 1199 | 92 | ||
More Information
Set Membership: |
|

Nr4a2
Genomic Map