Assay Details
| Gene Symbol: Hipk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145824.1 | NP_001139296.1 | 5 - 6 | 1977 | 99 | |
| NM_010434.2 | NP_034564.2 | 5 - 6 | 1977 | 99 | ||
| GenBank mRNA | AF077660.1 | - | 5 - 6 | 1731 | 99 | |
| AF170305.1 | - | 4 - 5 | 1441 | 99 | ||
| AF305238.1 | - | 4 - 5 | 1439 | 99 | ||
| AK140130.1 | - | 5 - 6 | 2080 | 99 | ||
| BC131663.1 | - | 5 - 6 | 1609 | 99 | ||
| BC131664.1 | - | 5 - 6 | 1608 | 40110751 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hipk3
Genomic Map