Assay Details
| Gene Symbol: Mki67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081117.2 | NP_001074586.2 | 8 - 9 | 2033 | 85 | |
| GenBank mRNA | AK052772.1 | - | 8 - 9 | 1953 | 85 | |
| AK082099.1 | - | 8 - 9 | 1952 | 85 | ||
| AK143677.1 | - | 8 - 9 | 2072 | 85 | ||
| X82786.1 | - | 8 - 9 | 2141 | 85 | ||
More Information
Set Membership: |
|

Mki67
Genomic Map