Assay Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253885.1 | NP_001240814.1 | 1 - 2 | 357 | 89 | |
| NM_001253886.1 | NP_001240815.1 | 1 - 2 | 357 | 89 | ||
| GenBank mRNA | AF206720.1 | - | 1 - 2 | 80 | 89 | |
| AK083830.1 | - | 1 - 2 | 87 | 89 | ||
| AK144340.1 | - | 1 - 2 | 358 | 89 | ||
| AK164140.1 | - | 1 - 2 | 82 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF206720.1,AK083830.1,AK164140.1,NM_001253885.1,NM_001253886.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Apbb1
Genomic Map