Assay Details
| Gene Symbol: Cckbr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF019371.1 | - | 2 - 3 | 402 | 63 | |
| AK028325.1 | - | 2 - 3 | 612 | 63 | ||
| AK039721.1 | - | 611 | 63 | |||
| AK053307.1 | - | 2 - 3 | 472 | 63 | ||
| BC103530.1 | - | 2 - 3 | 582 | 40039482 | 63 | |
| BC103531.1 | - | 2 - 3 | 582 | 40039474 | 63 | |
| BC103538.1 | - | 2 - 3 | 582 | 40039475 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cckbr
Genomic Map