Assay Details
| Gene Symbol: Cnga4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033317.3 | NP_001028489.1 | 4 - 5 | 1041 | 96 | |
| GenBank mRNA | AK136723.1 | - | 4 - 5 | 1043 | 96 | |
| BC107347.1 | - | 4 - 5 | 987 | 40055406 | 96 | |
| BC107348.2 | - | 3 - 4 | 1166 | 40055413 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cnga4
Genomic Map