Assay Details
| Gene Symbol: Fam160a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242363.2 | NP_001229292.1 | 5 - 6 | 1397 | 91 | |
| NM_001242364.2 | NP_001229293.1 | 4 - 5 | 1306 | 91 | ||
| NM_001242365.2 | NP_001229294.1 | 4 - 5 | 1306 | 91 | ||
| NM_199009.3 | NP_950174.1 | 4 - 5 | 1306 | 91 | ||
| GenBank mRNA | AK085350.1 | - | 4 - 5 | 1257 | 91 | |
| AK122552.1 | - | 4 - 5 | 1234 | 91 | ||
| AK154173.1 | - | 5 - 6 | 1341 | 91 | ||
| AK155273.1 | - | 5 - 6 | 1342 | 91 | ||
| AK159716.1 | - | 4 - 5 | 1261 | 91 | ||
| AK169996.1 | - | 5 - 6 | 1379 | 91 | ||
| BC018464.1 | - | 4 - 5 | 1223 | 91 | ||
| BC021532.1 | - | 2 - 3 | 633 | 91 | ||
| BC043717.1 | - | 3 - 4 | 822 | 91 | ||
| BC063093.1 | - | 4 - 5 | 1263 | 6332038 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fam160a2
Genomic Map