Assay Details
| Gene Symbol: Cd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009844.2 | NP_033974.2 | 14 - 15 | 1694 | 95 | |
| GenBank mRNA | AK089835.1 | - | 14 - 15 | 1693 | 95 | |
| AK156125.1 | - | 14 - 15 | 1694 | 95 | ||
| AK163608.1 | - | 12 - 13 | 2191 | 95 | ||
| AK172008.1 | - | 14 - 15 | 1656 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079079 | AK156125.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079079 | AK163608.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079079 | NM_009844.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2079079 | AK089835.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378223 | AK006482.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 378223 | BC048662.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 763440 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 223773 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 539432 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 582478 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P142325 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079079 | AK156125.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079079 | AK163608.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079079 | NM_009844.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2079079 | AK089835.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318966 | BE686865.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318966 | AK156125.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318966 | AK163608.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P318966 | NM_009844.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500365 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500371 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500362 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500379 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500378 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500359 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500367 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500383 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500382 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500363 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500376 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500354 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500353 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500374 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500398 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500380 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500361 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500370 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500369 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500392 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500360 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500355 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500352 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500357 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500388 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500390 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25500386 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P142325 | - | - |
| Whole Mouse Genome G4122A | A_51_P318966 | BE686865.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318966 | AK156125.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318966 | AK163608.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P318966 | NM_009844.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P129670 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121386 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P142325 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P05901085 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06443346 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06443343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500277 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500285 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500324 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500383 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500311 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500292 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500392 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500350 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500327 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500340 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500345 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500365 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500384 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500378 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500388 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500379 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500367 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500294 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500278 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500347 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500341 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500343 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500359 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500310 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500299 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500352 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500332 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500395 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500323 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500325 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500371 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500369 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500320 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500357 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500337 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500318 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500321 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500374 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500398 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500309 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500286 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500354 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500291 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500362 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500361 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500272 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500314 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500329 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500389 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500334 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500331 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500335 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500307 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500382 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500351 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500380 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25500390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500350 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06443343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500371 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987935 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05908959 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500382 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500313 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06342013 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500357 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500395 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500363 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500365 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500329 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500343 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500384 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500361 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06443346 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06152657 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500369 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500334 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500344 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500355 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500354 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500332 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500331 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500340 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500325 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500380 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500362 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06152659 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500341 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987925 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06410366 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500335 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987937 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500319 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500327 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05987936 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P05952123 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500390 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500338 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500347 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500377 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500337 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06150056 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500318 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500312 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500345 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500376 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25500360 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10567867 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567870 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567871 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567869 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567873 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567878 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567868 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567876 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567864 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567877 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567872 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567875 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567874 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567866 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10567865 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1450570_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10567863 | - | - |
| Mouse Genome 430 2.0 Array | 1450570_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1450570_PM_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12478_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 12478_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0700035632.mm.1 | NM_009844.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035632.mm.1 | NM_009844.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700035632.mm.1 | TC0700004277.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1450570_a_at | - | - |
| Mouse Expression Set 430 | 1450570_PM_a_at | - | - |
| Murine Genome U74v2 Set | 99945_at | - | - |
| Murine Genome U74v2 Set | 99945_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450570_a_at | - | - |
| Mouse Genome 430A 2.0 Array | 1450570_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450570_PM_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1450570_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 520081 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6940692 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 6940692 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940692 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228448 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1228448 | - | - |
| mouse-6 v1.1 (Illumina) | scl30689.14.1_1-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940692 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl30689.14.1_1-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940692 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2975994 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1228448 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2975994 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1228448 | - | - |

Cd19
Genomic Map