Assay Details
| Gene Symbol: Stap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145934.2 | NP_666046.1 | 11 - 12 | 1174 | 62 | |
| NR_102313.1 | 10 - 11 | 1278 | 62 | |||
| GenBank mRNA | AK050131.1 | - | 11 - 12 | 1154 | 62 | |
| AK050172.1 | - | 10 - 11 | 1246 | 62 | ||
| BC026642.1 | - | 11 - 12 | 1148 | 4986113 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stap2
Genomic Map