Assay Details
| Gene Symbol: Kank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030697.2 | NP_109622.1 | 4 - 5 | 1454 | 73 | |
| GenBank mRNA | AK002623.1 | - | 4 - 5 | 1455 | 73 | |
| AK132011.1 | - | 4 - 5 | 1476 | 73 | ||
| KJ650203.1 | - | 3 - 4 | 73 | |||
| KJ663713.1 | - | 3 - 4 | 73 | |||
| KJ663714.1 | - | 4 - 5 | 73 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Kank3
Genomic Map