Assay Details
| Gene Symbol: Daxx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199733.1 | NP_001186662.1 | 4 - 5 | 1545 | 56 | |
| NM_007829.4 | NP_031855.3 | 4 - 5 | 1445 | 56 | ||
| GenBank mRNA | AK133863.1 | - | 4 - 5 | 1353 | 56 | |
| AK144332.1 | - | 4 - 5 | 1377 | 56 | ||
| AK145906.1 | - | 4 - 5 | 1376 | 56 | ||
| AK146743.1 | - | 4 - 5 | 1372 | 56 | ||
| AK151279.1 | - | 4 - 5 | 1366 | 56 | ||
| AK153149.1 | - | 4 - 5 | 1333 | 56 | ||
| AK156425.1 | - | 4 - 5 | 1546 | 56 | ||
| AK172616.1 | - | 4 - 5 | 1549 | 56 | ||
| AY734459.1 | - | 2 - 3 | 1012 | 56 | ||
| AY734465.1 | - | 2 - 3 | 923 | 56 | ||
| AY734466.1 | - | 1203 | 56 | |||
| AY758273.1 | - | 2 - 3 | 927 | 56 | ||
| BC128373.1 | - | 4 - 5 | 1358 | 40111681 | 56 | |
| KJ650207.1 | - | 3 - 4 | 1275 | 56 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Daxx
Genomic Map