Assay Details
| Gene Symbol: Bet1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018742.5 | NP_061212.3 | 1 - 2 | 158 | 110 | |
| GenBank mRNA | AK158871.1 | - | 885 | 110 | ||
| AK161239.1 | - | 1 - 2 | 76 | 110 | ||
| BC019730.1 | - | 1 - 2 | 59 | 3667829 | 110 | |
| BY292666.1 | - | 1 - 2 | 159 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK158871.1, AK161239.1, BC019730.1, NM_018742.5 transcripts.
Set Membership: |
|

Bet1l
Genomic Map