Assay Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133946.2 | NP_598707.2 | 5 - 6 | 2329 | 67 | |
| GenBank mRNA | AK039531.1 | - | 2 - 3 | 1613 | 67 | |
| AY355343.1 | - | 2 - 3 | 1267 | 67 | ||
| BC013519.1 | - | 2 - 3 | 1236 | 67 | ||
| BC031139.1 | - | 2 - 3 | 1321 | 4972485 | 67 | |
| JF810536.1 | - | 4 - 5 | 2714 | 67 | ||
More Information
Set Membership: |
|

Nlrp6
Genomic Map