Assay Details
| Gene Symbol: Nlrp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133946.2 | NP_598707.2 | 6 - 7 | 2504 | 80 | |
| GenBank mRNA | AK039531.1 | - | 3 - 4 | 1788 | 80 | |
| AY355343.1 | - | 3 - 4 | 1442 | 80 | ||
| BC013519.1 | - | 3 - 4 | 1411 | 80 | ||
| BC031139.1 | - | 3 - 4 | 1496 | 4972485 | 80 | |
| FJ222825.1 | - | 6 - 7 | 2450 | 80 | ||
| JF810536.1 | - | 5 - 6 | 2889 | 80 | ||
More Information
Set Membership: |
|

Nlrp6
Genomic Map