Assay Details
| Gene Symbol: 2410137M14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029747.3 | NP_084023.1 | 3 - 4 | 462 | 84 | |
| GenBank mRNA | AK010800.1 | - | 3 - 4 | 463 | 84 | |
| BC147446.1 | - | 3 - 4 | 442 | 9056088 | 84 | |
| BC147465.1 | - | 3 - 4 | 442 | 9056107 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166660 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166663 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 874165 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 501562 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84761 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P116826 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166602 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166611 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166607 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166621 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166641 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166643 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166637 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166627 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166639 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166631 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166635 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166624 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166606 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166622 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166625 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166644 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166633 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166617 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166634 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166614 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166605 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166629 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166600 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166618 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166636 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31166603 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P116826 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08771965 | - | - |
| Whole Mouse Genome G4122A | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | NM_029747.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | AK010800.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC147446.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC147465.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P160397 | BC171992.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125330 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P116826 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524542 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524550 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524548 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524555 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13524544 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166663 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166661 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166662 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31166660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166596 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166604 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166576 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166564 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166660 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166580 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166657 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166578 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166573 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166633 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166539 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166639 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166563 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166644 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166646 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166591 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166631 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166655 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166624 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166629 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166651 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166567 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166607 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166618 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166667 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166601 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166570 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166586 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166588 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166627 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166653 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166530 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166641 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166533 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166611 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166614 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166585 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166658 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166531 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166647 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166592 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166581 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166594 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166648 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166621 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166534 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166649 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166535 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166590 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166617 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166537 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166625 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31166643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166600 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166598 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166627 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166576 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166637 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166618 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166585 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13524542 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166636 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166578 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166594 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166635 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166629 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13518160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166588 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166645 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166607 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166614 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13248453 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166570 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166572 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166591 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13529935 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166586 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166639 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13272398 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13110115 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166567 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166581 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542221 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166569 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13248456 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166579 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13524544 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13529936 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166590 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166573 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166609 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13352259 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13524549 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13504302 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166596 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166617 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166633 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166644 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166621 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31166575 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13542223 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10450838 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450839 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450836 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450837 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10450840 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1432454_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10450835 | - | - |
| Mouse Genome 430 2.0 Array | 1432454_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1432454_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76797_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 76797_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700019482.mm.1 | NM_029747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019482.mm.1 | NM_029747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019482.mm.1 | TC1700002045.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019485.mm.1 | NM_029747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019485.mm.1 | NM_029747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019485.mm.1 | TC1700002045.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700019483.mm.1 | NM_029747.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019483.mm.1 | NM_029747.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700019483.mm.1 | TC1700002045.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1432454_at | - | - |
| Mouse Expression Set 430 | 1432454_PM_at | - | - |
| Murine Genome U74v2 Set | 169523_at | - | - |
| Murine Genome U74v2 Set | 169523_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432454_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1432454_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 590743 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 3800563 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 3800563 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0003800563 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739645 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2739645 | - | - |
| mouse-6 v1.1 (Illumina) | scl49940.4.1_270-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 3800563 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl49940.4.1_270-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3800563 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2739645 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739645 | - | - |

2410137M14Rik
Genomic Map