Assay Details
Gene Symbol: H2-M2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008204.2 | NP_032230.1 | 3 - 4 | 627 | 132 | |
GenBank mRNA | AY302212.1 | - | 3 - 4 | 627 | 132 | |
AY302213.1 | - | 3 - 4 | 627 | 132 | ||
AY302217.1 | - | 3 - 4 | 627 | 132 | ||
BC150676.1 | - | 3 - 4 | 650 | 9087587 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302213.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302214.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302215.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302216.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P191463 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135064 | AY302217.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2135064 | NM_008204.2 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814318 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P139852 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135064 | AY302217.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2135064 | NM_008204.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168889 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168896 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168905 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168903 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168894 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168887 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168898 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168890 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168884 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168882 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168880 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168893 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168908 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168902 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168881 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168883 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168904 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168897 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168900 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168906 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168879 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168907 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168892 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168876 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168886 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168923 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31168901 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P139852 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101544 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P139852 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13150557 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13150562 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13150558 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13370799 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P13150567 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168906 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168901 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168905 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168883 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168920 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168881 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168916 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168899 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168894 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31168887 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168901 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168898 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168904 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168876 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168884 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168908 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168905 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168902 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168892 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13370799 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168871 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168894 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168870 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13150559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168880 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168879 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168890 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168874 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13244823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13150560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13065838 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168881 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13065841 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168897 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13150567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31168906 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10450886 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450884 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450885 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450883 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450887 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450881 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10450882 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10450880 | - | - |
Affymetrix Mouse Clariom S Array | PSR1700019573.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019573.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019573.mm.1 | TC1700002071.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019574.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019574.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019574.mm.1 | TC1700002071.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019575.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019575.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019575.mm.1 | TC1700002071.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019579.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019579.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019579.mm.1 | TC1700002071.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019576.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019576.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019576.mm.1 | TC1700002071.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1700019578.mm.1 | NM_008204.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019578.mm.1 | NM_008204.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1700019578.mm.1 | TC1700002071.mm.1 | MTA TC |
Murine Genome U74v2 Set | 101300_PM_at | - | - |
Murine Genome U74v2 Set | 101300_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 661903 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseRef-8 v2 Expression BeadChip | ILMN_2964185 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2964185 | - | - |
