Assay Details
| Gene Symbol: Brsk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009929.3 | NP_001009929.1 | 8 - 9 | 1119 | 88 | |
| NM_001009930.3 | NP_001009930.1 | 8 - 9 | 1119 | 88 | ||
| NM_001276763.1 | NP_001263692.1 | 8 - 9 | 1119 | 88 | ||
| NM_029426.2 | NP_083702.1 | 8 - 9 | 1119 | 88 | ||
| GenBank mRNA | AK158574.1 | - | 8 - 9 | 868 | 88 | |
| AK173268.1 | - | 7 - 8 | 694 | 88 | ||
| AY533672.1 | - | 8 - 9 | 784 | 88 | ||
| AY533673.1 | - | 8 - 9 | 784 | 88 | ||
| AY533674.1 | - | 8 - 9 | 784 | 88 | ||
| AY660739.1 | - | 8 - 9 | 1119 | 88 | ||
| BC056498.1 | - | 5 - 6 | 395 | 88 | ||
| JF707772.1 | - | 8 - 9 | 784 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Brsk2
Genomic Map