Assay Details
| Gene Symbol: Acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025826.4 | NP_080102.1 | 8 - 9 | 1133 | 106 | |
| GenBank mRNA | AK004889.1 | - | 8 - 9 | 1034 | 106 | |
| AK076535.1 | - | 8 - 9 | 1034 | 106 | ||
| AK152325.1 | - | 8 - 9 | 1022 | 106 | ||
| AK163322.1 | - | 4 - 5 | 614 | 106 | ||
| BC011235.1 | - | 8 - 9 | 1015 | 106 | ||
| BC035231.1 | - | 8 - 9 | 999 | 106 | ||
| BC044826.1 | - | 8 - 9 | 1066 | 106 | ||
| BC054428.1 | - | 8 - 9 | 1075 | 5039627 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Acadsb
Genomic Map