Assay Details
| Gene Symbol: Chst15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029935.5 | NP_084211.2 | 6 - 7 | 1912 | 61 | |
| GenBank mRNA | AB187269.1 | - | 5 - 6 | 1352 | 61 | |
| AF320786.1 | - | 5 - 6 | 1403 | 61 | ||
| AK019474.1 | - | 6 - 7 | 1867 | 61 | ||
| AK031745.1 | - | 4 - 5 | 613 | 61 | ||
| AK035637.1 | - | 6 - 7 | 2000 | 61 | ||
| AK043803.1 | - | 4 - 5 | 760 | 61 | ||
| AK082318.1 | - | 6 - 7 | 1978 | 61 | ||
| AK083614.1 | - | 6 - 7 | 1912 | 61 | ||
| AK122324.1 | - | 6 - 7 | 2213 | 61 | ||
| BC031443.1 | - | 6 - 7 | 1869 | 4240286 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chst15
Genomic Map