Assay Details
| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007865.3 | NP_031891.2 | 3 - 4 | 995 | 60 | |
| GenBank mRNA | BC057400.1 | - | 3 - 4 | 508 | 6402691 | 60 |
| BC065063.1 | - | 3 - 4 | 759 | 6416191 | 60 | |
| X80903.1 | - | 3 - 4 | 425 | 60 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dll1
Genomic Map