Assay Details
| Gene Symbol: Dll1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007865.3 | NP_031891.2 | 4 - 5 | 1245 | 77 | |
| GenBank mRNA | BC057400.1 | - | 4 - 5 | 758 | 6402691 | 77 |
| BC065063.1 | - | 4 - 5 | 1009 | 6416191 | 77 | |
| X80903.1 | - | 4 - 5 | 675 | 77 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dll1
Genomic Map