Assay Details
| Gene Symbol: Tbp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC012685.1 | - | 2 - 3 | 328 | 3498542 | 113 |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC012685.1 transcripts.
Set Membership: |
|

Tbp
Genomic Map