Assay Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110794.1 | NP_001104264.1 | 5 - 6 | 666 | 62 | |
| NM_009674.3 | NP_033804.2 | 5 - 6 | 781 | 62 | ||
| GenBank mRNA | AK032013.1 | - | 5 - 6 | 557 | 62 | |
| AK077579.1 | - | 5 - 6 | 474 | 62 | ||
| AK149654.1 | - | 5 - 6 | 473 | 62 | ||
| AK151438.1 | - | 5 - 6 | 475 | 62 | ||
| AK161613.1 | - | 5 - 6 | 475 | 62 | ||
| AK169199.1 | - | 5 - 6 | 448 | 62 | ||
| AK178561.1 | - | 2 - 3 | 62 | |||
| BC008997.1 | - | 5 - 6 | 3157926 | 62 | ||
| L13129.1 | - | 5 - 6 | 468 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa7
Genomic Map