Assay Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110794.1 | NP_001104264.1 | 6 - 7 | 771 | 71 | |
| NM_009674.3 | NP_033804.2 | 6 - 7 | 886 | 71 | ||
| GenBank mRNA | AK032013.1 | - | 6 - 7 | 662 | 71 | |
| AK077579.1 | - | 6 - 7 | 579 | 71 | ||
| AK134589.1 | - | 7 - 8 | 656 | 71 | ||
| AK151438.1 | - | 6 - 7 | 580 | 71 | ||
| AK161613.1 | - | 6 - 7 | 580 | 71 | ||
| AK167588.1 | - | 6 - 7 | 554 | 71 | ||
| AK169199.1 | - | 6 - 7 | 553 | 71 | ||
| BC008997.1 | - | 6 - 7 | 570 | 3157926 | 71 | |
| L13129.1 | - | 6 - 7 | 573 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Anxa7
Genomic Map